Learn More
tetrahydro-2h-pyran-4-ylmethanol, 97%, Thermo Scientific™
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
Specifications
Assay Percent Range | 97% |
Packaging | Amber Glass Bottle |
Quantity | 250 mg |
IUPAC Name | oxan-4-ylmethanol |
Formula Weight | 116.16 |
Percent Purity | 97% |
Chemical Name or Material | Tetrahydro-2H-pyran-4-ylmethanol |