missing translation for 'onlineSavingsMsg'
Learn More

Tetradecyl sodium sulfate, 95%

Product Code. p-4228911 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 10458862

Brand: Thermo Scientific Acros 420260050

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 1191-50-0 | C14H29NaO4S | 316.43 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1191-50-0
Molecular Formula C14H29NaO4S
Molecular Weight (g/mol) 316.43
MDL Number MFCD00007468
InChI Key UPUIQOIQVMNQAP-UHFFFAOYSA-M
Synonym sodium tetradecyl sulfate, sodium myristyl sulfate, niaproof 4, sodium tetradecyl sulphate, tesapon k 14, texapon k 14, trombavar, natri tetradecylsulfas, sotradecol sodium, tetradecyl sodium sulfate
PubChem CID 23665770
IUPAC Name sodium;tetradecyl sulfate
SMILES [Na+].CCCCCCCCCCCCCCOS([O-])(=O)=O

Specifications

Melting Point 199.0°C
Color White
Elemental Analysis 94% min.
Infrared Spectrum Authentic
Assay Percent Range 94% min
Packaging Glass bottle
Linear Formula CH3(CH2)13OSO3Na
Quantity 5 g
Formula Weight 316.44
Percent Purity 95%
Physical Form Micro-crystalline Powder
Chemical Name or Material Tetradecyl sodium sulfate
Show More Show Less
compliance-icons
Product Identifier
  • Sodium tetradecyl sulphate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Supplemental information
  • MIXTURE LIST-Contains : Sodium tetradecyl sulphate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.