missing translation for 'onlineSavingsMsg'
Learn More

TES, 99%, Thermo Scientific Chemicals

Catalog Number p-7026043
Click to view available options
Quantity:
10 g
50 g
250 g
This item is not returnable. View return policy
7365-44-8
C6H15NO6S
229.247
MFCD00007532
JOCBASBOOFNAJA-UHFFFAOYSA-N
tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine
81831
CHEBI:44356
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
C(CS(=O)(=O)O)NC(CO)(CO)CO
This item is not returnable. View return policy
7365-44-8
C6H15NO6S
229.247
MFCD00007532
JOCBASBOOFNAJA-UHFFFAOYSA-N
tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine
81831
CHEBI:44356
2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
C(CS(=O)(=O)O)NC(CO)(CO)CO

TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TES plays an important role in production of pH buffers for a biological system. It is used to make zwitterionic buffer solutions in the pH range 6.8-8.2. It is used in molecular biology, diagnosis, cell culture, pharmacy, agrochemical and pharmaceuticals. It is also used in the study of succinate oxidation and one of the components of test yolk buffer medium used in refrigeration of semen and transport.

Solubility
Soluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Melting Point ∼225°C (decomposition)
Odor Odorless
Linear Formula (HOCH2)3CNHCH2CH2SO3H
Quantity 10 g
Beilstein 1957061
Merck Index 14,9176
Solubility Information Soluble in water.
Formula Weight 229.25
Percent Purity 99%
Chemical Name or Material TES

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'