Learn More
TentaGel™ S-RAM, Thermo Scientific Chemicals
Brand: Thermo Scientific Acros 354650050
Additional Details : Weight : 0.00500kg
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
For solid support synthesisSpecifications
White to Yellow | |
Powder | |
Glass bottle | |
5 g | |
N-FMOC-4'-[poly(oxyethylene)carbamoylmethoxy]-2, 4-dimethoxy-benzhydrylamine polymer bound, Poly(oxyethylene)-RAM polymer bound |
TentaGel™ S-RAM | |
0.2 to 0.3mmol/g | |
90 Micron | |
MFCD00217896 |
RUO – Research Use Only