Learn More
Fisherbrand™ Temperature Strip
Brand:  Fisherbrand™
Additional Details : Weight : 0.00180kg
 View more versions of this product
Product Code. 11512513
  • POA / Pack of 10
Estimated Shipment: 19-06-2024
to see stock.
This item requires a price update prior to being ordered.
Request a Price for this item here.
Description
Description
The standard 8 level range covers 40°C to 260°C for general purpose temperature monitoring.
Specifications
Specifications
204°C
166°C to 204°C
5°C
166°C
0.02