Learn More
TMC Hallcrest™ Thermax™ 5 Level Clock Temperature Indicators
Provide a fast, simple temperature monitoring system based on proprietary color change technology. TMC Hallcrest™ Thermax™ 5 Level Clock Temperature Indicators record the highest temperature achieved with irreversible color change.
Brand: TMC Hallcrest™ M05CTHERG4D01P1
Additional Details : Weight : 0.00450kg
Description
- Heat-sensitive round label to monitor sterilization temperature
- Inexpensive and easy to use
- Adhesive
- Immediate response.
- Permanent color change records highest temperature achieved.
- Oil, water and steam resistant.
- Accuracy tolerances: 29-99°C (+/-1°C) / 100 -154°C (+/-1.5°C) 160-290°C (+/-1°C)
Specifications
116°C to 138°C | |
116/121/127/132/138°C | |
116°C | |
14 mm | |
Polyester Film, Acrylic Adhesive |
±1°C | |
138°C | |
0.15 in. | |
Yes | |
Circular Temperature Measurement Indicators |