Learn More
Ebro Electronic™ TDC 150 Thermometer
Thermometer, °C / °F-switchable
Brand: Ebro Electronic™ 1340-1611
Additional Details : Weight : 0.10000kg
Description
Therm. (Needle length: 120 mm, dia. 3mm), pointed, tef.cable 1.5mSpecifications
EMC-Directive 2014/30/EU; EN 61326-1:2013; RoHS Directive 2011/65/EU; Electromagnetic Compatibility Regulations 2016 BS EN 61326-1:2013; The Restriction of the Use of Certain Hazardous Substances in Electrical and Electronic Equipment Regulations 2012 | |
7 mm | |
Applications: Temperature measurement, laboratory, process monitoring, | |
180 mm | |
TDC 150 | |
0.1°C | |
Low Cost Thermometer | |
302°F | |
-58°F | |
-50°C to +150°C | |
Approx. 5,000 hours | |
±1°C | |
LCD-7mm Display |
3.5 mm | |
Single-use | |
24 mm | |
125 mm | |
Stainless Steel Pointed | |
0.08 kg | |
None | |
150°C | |
-50°C | |
No | |
Lithium LR44 | |
60 mm | |
100 mm |