Learn More
Tartronic acid, 97%
Click to view available options
Quantity:
0.1 g
0.5 g
Unit Size:
0.1g
0.5g
Beskrivning
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.
Solubility
Soluble in water and alcohol.
Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
Tartronic acid is used as a reactant in the catalytic oxidation with air to form mesoxalic acid.
Solubility
Soluble in water and alcohol.
Notes
Store in a cool place. Incompatible with bases, oxidizing agents and reducing agents.
Specifikationer
Specifikationer
| Melting Point | ∽151°C (decomposition) |
| Assay Percent Range | 0.98 |
| Beilstein | 1209791 |
| Merck Index | 14,9073 |
| Solubility Information | Soluble in water and alcohol. |
| Formula Weight | 120.06 |
| Percent Purity | 98% |
| Quantity | 0.1 g |
| Chemical Name or Material | Tartronic acid |
RUO – Research Use Only
Korrigering av produktinnehåll
Din input är viktig för oss. Fyll i det här formuläret för att ge feedback relaterad till innehållet på denna produkt.
Produkttitel
Hittar du en möjlighet till förbättring?Dela en innehållskorrigering