Learn More
MP Biomedicals™ Taq Core Kit 25
Taq DNA Polymerase + dNTPs in one kit
578.76 EUR
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
11477030
|
MP Biomedicals™
11EPTQK253 |
3 x 250 u |
578.76 EUR
750 units |
Estimated Shipment: 29-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
- Taq CORE Kits contain all the reagents (presented as separate items) required for PCR:
- MP Biomedicals high quality recombinant Taq DNA polymerase, high purity dNTPs Mix (>99%), optimized incubation buffers with and without MgCl2 and a separate MgCl2 solution.
- dNTPs at 25mM each