missing translation for 'onlineSavingsMsg'
Learn More

TAPS sodium salt, 98%, Thermo Scientific Chemicals

Catalog Number p-7042047
Click to view available options
Quantity:
25 g
100 g
This item is not returnable. View return policy
C7H16NNaO6S
265.256
MFCD00069903
FEGYIWVHCSRXCG-UHFFFAOYSA-M
23667519
sodium;3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonate
C(CNC(CO)(CO)CO)CS(=O)(=O)[O-].[Na+]
This item is not returnable. View return policy
C7H16NNaO6S
265.256
MFCD00069903
FEGYIWVHCSRXCG-UHFFFAOYSA-M
23667519
sodium;3-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]propane-1-sulfonate
C(CNC(CO)(CO)CO)CS(=O)(=O)[O-].[Na+]

Zwitterionic buffer

TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
TAPS sodium salt is used in biochemistry and molecular biology. It has a useful pH range of 7.7 - 9.1. It has been utilized in capillary electrophoresis of such materials as DNA and DNA-dye complexes. Dyes have been separated by planar chromatography with electro osmotic flow with 1 mM TAPS in the mobile phase. The stability constants of metal ions with TAPS have been investigated by capillary electrophoresis. It has also been shown to inhibit connexin channel activity. The activity of various mutants of carbonic anhydrase I and carbonic anhydrase II has been studied in a variety of buffers, including TAPS.

Solubility
Soluble in water (333.33 mg/ml).

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY
Color White
Physical Form Powder
Quantity 25 g
Linear Formula (HOCH2)3CNH(CH2)3SO3Na
Solubility Information Soluble in water (333.33mg/ml).
Formula Weight 265.26
Percent Purity 98%
Chemical Name or Material TAPS sodium salt

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'