missing translation for 'onlineSavingsMsg'
Learn More
Learn More
DWK Life Sciences Tamper Evident Screw Cap, Wide Mouth, Blue PP, for Soda-Lime Glass Screw-Top Square Bottles
14.35 EUR valid until 2024-12-31
Use promo code "23258" to get your promotional price.
Use promo code "23258" to get your promotional price.
Click to view available options
Thread Size (Metric):
GL 32
GL 45
GL 54
GL 60
Description
Tamper evident, blue polypropylene, linerless screw cap for wide mouth/short form DIN thread soda lime glass bottles.
- Tamper evident ring
- LInerless
- Short form DIN thread
Specifications
Diameter (Metric) | 58 mm |
For Use With (Application) | Storage of samples and reagents; Sample collection; Transport and shipping |
Material | Polypropylene |
Quantity | 10/Pk. |
Thread Size (Metric) | GL 45 |
Autoclavable | Autoclavable |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'