missing translation for 'onlineSavingsMsg'
Learn More

Brady™ Lockout Labels

Brand:  Brady™ 256201

Additional Details : Weight : 0.05000kg

 View more versions of this product

Product Code. 11766865

  • 25.65 EUR / 1 pack
Estimated Shipment: 17-05-2024
to see stock.
Add to basket

Description

Description

Specifications

Specifications

10 Pk.
Red on White
Ne pas mettre en marche. Ne pas enlever cette condamnation
6.3 in.
0.249 mm
-40°F to +212°F
75 mm
7 mm
Tag
Polyester
Lockout Tagout
160 mm
-40°C to +100°C
2.95 in.
160 x 75 mm
Lockout Tagout Tags
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers