Learn More
MedChemExpress T7 Tag Peptide
Product Code. 16442789
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16442789 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
Artikelnummer. 16442789 Lieferant MedChemExpress Lieferanten-Nr. HYP0327
This item requires a price update prior to being ordered.
Request a Price for this item here.
Dieser Artikel kann nicht zurückgegeben werden. Rückgaberichtlinie anzeigen
MedChemExpress T7 Tag Peptide is a protein tag derived from the N-terminal 11 residues of the major T7 capsid protein, gp 10. T7 Tag Peptide can be used in different immunoassays as well as affinity purification.
Spezifikation
CAS 245445-88-9
Content And Storage Please store the product under the recommended conditions in the Certificate of Analysis.
Molecular Weight (g/mol) 1098.27 g/mol
Product Type Peptides
Quantity 1 mg
Shipping Condition Room temperature