Learn More
Eppendorf™ Swing Bucket Rotors
Perfect for phase-separation applications such as phenol-chloroform extraction
Brand:  Eppendorf™ 5492008003
Additional Details : Weight : 1.33000kg
 View more versions of this product
Product Code. 10039220
  • 663.00 EUR / Each
Estimated Shipment
12-05-2021
Description
Description
  • Designed for use with Eppendorf Centrifuges
Specifications
Specifications
Swinging Bucket Rotor
10,600 x g
5417R and 5417C Microcentrifuges
8 x 1.5/2mL
10,000rpm
Aluminum lid