Learn More
3M™ Swab-Sampler
Convenient and quick collection device for environmental sampling and testing over small surface areas.
Brand:  3M™ 7000030301
Additional Details : CAS Number : 7732-18-5 Weight : 0.48500kg
 View more versions of this product
Product Code. 17248520
  • 169.60 EUR / Pack of 100
Estimated Shipment: 26-06-2024
Description
  • Letheen Broth vial is ready to use for pathogen testing of environmental samples.
  • Swab reaches into corners and hard-to-reach areas for increased accuracy.
  • Convenient design helps improve sampling consistency from tech-to-tech and plant-to-plant.
  • Bottles have ample headspace for sample addition and vortexing.
  • Self-contained swab and vial for increased productivity and efficiency.
Specifications
Specifications
4 mL Neutralizing Buffer
100 /Cs.
Swab