Learn More
Thermo Scientific™ Storerooms
Organize supplies, save valuable bench space using these six or nine drawer Storerooms. Order an empty cabinet or one fully stocked with vials and closures.
Brand:  Thermo Scientific™ C5075-213
Additional Details : Weight : 0.20000kg
 View more versions of this product
Product Code. 15260379
  • 850.00 EUR / Each
Estimated Shipment
07-08-2019
Add to basket
Specifications
Specifications
Nine-Drawer Storeroom, Caps and Clear Glass Vials with ID Patch
Yes
Holds 2000 caps and glass vials with ID patch
Glass
Caps and Clear Glass Vials
1
Clear
Videos
Documents
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.