missing translation for 'onlineSavingsMsg'
Learn More
Learn More
STXBP5 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 24512-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
Syntaxin 1 is a component of the 7S and 20S SNARE complexes which are involved in docking and fusion of synaptic vesicles with the presynaptic plasma membrane. This gene encodes a syntaxin 1 binding protein. In rat, a similar protein dissociates syntaxin 1 from the Munc18/n-Sec1/rbSec1 complex to form a 10S complex, an intermediate which can be converted to the 7S SNARE complex. Thus this protein is thought to be involved in neurotransmitter release by stimulating SNARE complex formation. Alternatively spliced transcript variants encoding different isoforms have been identified.Specifications
STXBP5 | |
Polyclonal | |
Unconjugated | |
STXBP5 | |
LGL3, LLGL3, Nbla04300, STXBP5, Syntaxin binding protein 5, Tomosyn 1 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
134957, 81022 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Immunoprecipitation, Western Blot | |
0.3 mg/mL | |
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
Q5T5C0, Q9WU70 | |
Stxbp5 | |
STXBP5 Fusion Protein Ag21629 | |
150 μL | |
Primary | |
Human, Rat | |
Antibody | |
IgG |