Learn More
Techne™ Dri-Block™ Aluminum Heating Block Insert
Click to view available options
96-well x 1 mL plate, no thermometer hole
Description
- Versatile range of interchangeable heating blocks work with many size sample tubes and plates
- Heaters sold separately
Specifications
Description | No thermometer hole |
Material | Anodized Aluminum |
No. of Places | 96 |
Type | Dri-Block™ Insert |
Diameter (Metric) Well | 1 mL |