missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ eBioscience™ Streptavidin eFluor™ 660 Conjugate
Commonly used in indirect staining protocols to detect biotinylated primary antibodies in flow cytometry, immunohistochemistry, and immunocytochemistry applications. Streptavidin binds to biotin with high affinity.
172.00 EUR - 438.00 EUR
Specifications
Concentration | 0.2 mg/mL |
---|---|
Conjugate | eFluor 660 |
Alert:
Contains Sodium azide
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15391900
|
Invitrogen™
50-4317-82 |
100 μg |
438.00 EUR
100µg |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
15381900
|
Invitrogen™
50-4317-80 |
25 μg |
172.00 EUR
25µg |
Estimated Shipment: 29-04-2024
Log in to see stock. |
|||||
Specifications
0.2 mg/mL | |
eFluor 660 |
For Research Use Only