Learn More
Molecular Probes™ Streptavidin, Alexa Fluor™ 680 Conjugate
Streptavidin, Alexa Fluor™ 680 conjugate
409.00 EUR - 473.00 EUR
Specifications
Conjugate | Alexa Fluor 680 |
---|---|
Product Line | Alexa Fluor™ |
Product Type | Streptavidin Conjugate (fluorescent) |
Label or Dye | Alexa Fluor Dyes |
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Form | Quantity | Price | Quantity & Availability | ||||
10750574
|
Molecular Probes™
S21378 |
Solid | 1 mg |
409.00 EUR
1mg |
In Stock
Log in to see stock. |
||||
10359222
|
Molecular Probes™
S32358 |
Liquid | 0.5 mL |
473.00 EUR
Each |
In Stock
Log in to see stock. |
||||
Description
Alexa Fluor™ 680 streptavidin comprises a biotin-binding protein (streptavidin) covalently attached to a fluorescent label (Alexa Fluor™ dye). Streptavidin has a very high binding affinity for biotin, and a conjugate of streptavidin is commonly used together with a conjugate of biotin for specific detection of a variety of proteins, protein motifs, nucleic acids, and other molecules (for example, a biotinylated primary antibody bound to a protein target can be detected with a fluorescently labeled streptavidin). Strategies similar to this are used in many detection protocols including western blots, flow cytometry, imaging and microscopy, and microplate assays. Alexa Fluor™ dye streptavidin conjugates are supplied as 1 mg lyophilized product or in 0.5 mL volumes of a 2 mg/mL solution.Important Features of Alexa Fluor™ 680 Streptavidin Conjugates:
• Alexa Fluor™ 680 streptavidin conjugate has Ex/Em maxima of ∼ (679/702)
• Bright, photostable fluorescence
• High solubility in aqueous solutions
• Available in multiple colors
• Ideal for western blots, flow cytometry, imaging and microscopy, microplate assays and more
Properties of Alexa Fluor™ Dyes
Alexa Fluor™ dyes are organic fluorescent dyes developed for better performance in imaging and other labeling protocols and exhibit improved photostability and brightness and improved solubility in aqueous solutions. Available in a broad range of colors, these dyes are a good choice for most imaging applications.
Blocking Endogenous Biotin
Naturally occurring biotins can interfere with biotin-streptavidin detection schemes. For experiments involving fixed and permeabilized cells, try our Endogenous Biotin-Blocking Kit to minimize this interference.
For Research Use Only. Not intended for any animal or human therapeutic or diagnostic use.
Specifications
Alexa Fluor 680 | |
Alexa Fluor™ | |
Streptavidin Conjugate (fluorescent) | |
Alexa Fluor Dyes |
For Research Use Only. Not for use in diagnostic procedures.