Learn More
IBA Lifesciences StrepMan Magnet
IBA Lifesciences GmbH StrepMan Magnet is designed to be used for manual cell isolation with Strep-Tactin™ Magnetic Microbeads using the Streptamer™ Technology.
Brand:  IBA Lifesciences 6-5650-065
Additional Details : Weight : 0.00900kg
Product Code. 15362096
  • 386.00 EUR / Each
Estimated Shipment: 16-05-2024
Description
  • The StrepMan consists of a plastic frame with two embedded permanent magnets and accommodates two different tube sizes.
  • It is fit for 2x 15 ml and 2x 50 ml conical reaction tubes.
  • It can be used for magnetic cell isolation with microbeads such as Strep-Tactin™ Magnetic Microbeads.
Specifications
Specifications
For 2 x 15 mL and 2 x 50 mL Conical Reaction Tubes