Learn More
Lenz Laborglasintrumente Stoppers Octagonal Plate, Glass
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand: Lenz Laborglasintrumente 01400545
This item is not returnable.
View return policy
Description
- Acc. to DIN 12252
- Form A and E
- Solid or semi-hollow
- Material: glass
Specifications
Semi-hollow | |
Borosilicate Glass |