Learn More
Lenz Laborglasintrumente Stoppers Octagonal Plate, Glass
Made of high quality materials. Each manufacturing step is tested through an extensive quality management system, based on DIN ISO 9001/EN 29001.
Brand:  Lenz Laborglasintrumente 01400545
 View more versions of this product
Product Code. 11866134
  • 26.98 EUR / Each
Estimated Shipment: 25-11-2024
to see stock.
Explore more special offers
Description
  • Acc. to DIN 12252
  • Form A and E
  • Solid or semi-hollow
  • Material: glass
Specifications
Specifications
Semi-hollow
Borosilicate Glass