Learn More
Corning™ Sterile Plastic Octagonal Media Bottle with Cap
Corning™ plastic octagonal media bottle with screw cap used for sampling and storing
Brand:  Corning™ PURV25003
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 11744965
  • 758.00 EUR / Pack of 144
Estimated Shipment
03-08-2021
Add to basket
Description
Description
  • Features:
    • Sterilized to SAL 10-6, making it suitable for immediate use
    • Screw cap is tamper-evident and prevents unnecessary contamination
    Uses:
    • For sampling and storing media, biologicals, and other types of aqueous solutions, commonly used in labs
Specifications
Specifications
Yes
PET
-40°C to +50°C
50 to 250 mL (at 50 mL intervals)
250 mL
Sterile
31.7 mm