Learn More
First Aid Kit Sterile Dressing Refill
Keep a First Aid Kit Sterile Dressing Refill on hand per the Health and Safety ACOP L74 regulation for contingency supplies in case of emergency.
7.30 EUR - 10.45 EUR
Specifications
Product Type | Dressing |
---|---|
Sterility | Sterile |
Quantity | 6 Pk. |
For Use With (Application) | For first aid kit |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
12910465
|
Safety First
D7632T/ |
Large |
10.45 EUR
Pack of 6 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
12900465
|
Safety First
D7631T/ |
Medium |
7.30 EUR
Pack of 6 |
Estimated Shipment: 19-06-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
With reference to first aid kit refills, the Health and Safety ACOP L74 states “Sufficient supplies should be held as back-up on site”.Specifications
Dressing | |
Sterile | |
6 Pk. | |
For first aid kit |