missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tenak™ Standard Rack for Microtiterplates, for Chest Freezers, 16 mm Shelf Height
Stainlesss Steel Standard Rack for chest freezers is designed to hold microtiterplates.
113.00 EUR - 227.00 EUR
Specifications
Includes | Locking rod |
---|---|
Material | Stainless Steel |
For Use With (Equipment) | Chest Frseezers |
Height (Metric) | 16 mm |
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Holds | Price | Quantity & Availability | |||||
15200874
|
Tenak™
TE27102 |
Microtiterplates (19) |
125.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
15210874
|
Tenak™
TE27104 |
Microtiterplates (22) |
137.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
15220874
|
Tenak™
TE27106 |
Microtiterplates (26) |
149.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
15230874
|
Tenak™
TE27108 |
Microtiterplates (29) |
161.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
15240874
|
Tenak™
TE27110 |
Microtiterplates (32) |
173.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
15250874
|
Tenak™
TE27112 |
Microtiterplates (35) |
227.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
15260874
|
Tenak™
TE27114 |
Microtiterplates (39) |
197.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
15270874
|
Tenak™
TE27116 |
Microtiterplates (42) |
209.00 EUR
Each |
Estimated Shipment: 16-07-2024
Log in to see stock. |
|||||
15290864
|
Tenak™
TE27100 |
Microtiterplates (16) |
113.00 EUR
Each |
Estimated Shipment: 17-07-2024
Log in to see stock. |
|||||
Description
- For use in chest freezers
- Racks come with a locking rod to secure the cryoboxes
- Constructed of stainless steel
Specifications
Locking rod | |
Stainless Steel | |
Chest Frseezers | |
16 mm |