Learn More
RETSCH Stainless-Steel Test Sieves, 315mm Dia., in Pore Size 10mm and Smaller
Brand:  RETSCH 60.172.005000
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11712976
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
Specifications
Stainless Steel
Stainless Steel
1 Each
Test Sieve
For screening aggregates, separation, fractioning, particle size determination
315 mm
5mm
ISO 3310 and ASTM E 11, DIN 3310/2