Learn More
RETSCH Stainless-Steel Test Sieves, 315mm Dia., in Pore Size 10mm and Smaller
Brand: RETSCH 60.172.002360
Additional Details : Weight : 0.80000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve |
For screening aggregates, separation, fractioning, particle size determination | |
315 mm | |
2.36mm | |
ISO 3310 and ASTM E 11, DIN 3310/2 |