Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 50mmH, with Perforated Plates with Round Holes and Pore Size 10mm and Smaller
Brand: RETSCH 60.198.001400
216.60 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 1.00000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve | |
Certificate indicating Compliance |
For separation, fractioning, particle size determination | |
200 mm | |
1.4mm | |
ISO 3310/2 | |
50mm |