Learn More
RETSCH Stainless-Steel Test Sieves, 200 Dia. x 25mmH, in Micrometer Pore Sizes with ASTM E 11
Brand: RETSCH 60.123.000180
116.85 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.20000kg
Specifications
Stainless Steel | |
Stainless Steel | |
1 Each | |
Test Sieve | |
Shaker AS 200 | |
25mm |
For separation, fractioning, particle size determination | |
200 mm | |
180μm | |
ASTM E 11 | |
Certificate indicating Compliance |