Learn More
Cole-Parmer™ Stainless Steel Propellers
Use this four-blade propeller for general-purpose vortex mixing. Cole-Parmer&trade Stainless Steel Propellers are ideal for a variety of applications.
Brand: Cole-Parmer™ 04370-13
Additional Details : Weight : 0.80000kg
Description
- Stainless steel four-blade propeller
- Threaded 10 mm dia. bore
- Fits only shaft model 04370-02
Specifications
40 mm | |
1 Each | |
Propeller |