Learn More
Agencinox™ Stainless Steel Flat Bottom Dish
Use these dishes in decontamination services, operating rooms, surgeries, clinics and healthcare fields. Agencinox™ Stainless Steel Flat Bottom Dishes are ideal for holding dressings and other items which may be needed during surgical procedures.
15.74 EUR - 51.27 EUR
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Price | Quantity & Availability | |||||
11729503
|
Agencinox™
304 |
20 mL |
51.27 EUR
Pack of 5 |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11847503
|
Agencinox™
310A |
250 mL |
42.45 EUR
Each |
Estimated Shipment: 14-06-2024
Log in to see stock. |
|||||
11776533
|
Agencinox™
312 |
500 mL |
15.74 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
11786533
|
Agencinox™
316 |
1000 mL |
31.25 EUR
Each |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- Flat bottom stainless steel lotion bowls
- Autoclavable
- Available in a variety of sizes
Specifications
Flat | |
Stainless Steel | |
Round | |
Dish |