Learn More
RETSCH Stainless Steel Bottom Sieve For SM 200 Cutting Mills
Brand:  RETSCH 03.647.0320
288.80 EUR valid until 2024-12-31
Use promo code "23265" to get your promotional price.
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11700765
  • 304.00 EUR / Each
Estimated Shipment: 11-06-2024
Specifications
1Each
Bottom Sieves
SM 200 Cutting Mill
4mm