Learn More
Masterflex™ Stainless Steel Barbed Fitting NPT Male Pipe Adapter
Designed for quick and easy assembly without tools
Brand: Masterflex™ 31210-61
Additional Details : Weight : 0.80000kg
Description
- Available in a variety of materials and configurations to meet your need
- Adapts from a NPT male thread to a hose barb
Specifications
Barbed Fitting NPT Male Pipe Adapter | |
Stainless Steel | |
1.5 in. | |
1.5 in. NPT(M) | |
1.5 in. NPT(M) |
1 1/2 NPT(M) x 1 1/2 in. Tubing ID | |
3 7/8 in. | |
1.5 in. | |
1.5 in. | |
2 in. |