Learn More
Hanna Instruments™ Stainless Steel ATC Probes
AISI 316 stainless steel temperature probe that allows for temperature compensated measurements on compatible Hanna meters in laboratory environments.
Brand: Hanna Instruments™ HI7669/2
Additional Details : Weight : 1.00000kg
Description
- Stainless steel temperature probe with a 1 m cable length that allows for temperature compensated measurements on compatible Hanna meters
- Stainless steel temperature probe that can be used for compatible Hanna meters
- AISI 316 stainless steel
- NTC thermistor
- Wide measurement range
Specifications
ATC Probe | |
Stainless Steel |