missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SQRDL Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17256-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
SQRDL (Sulfide: quinone oxidoreductase, mitochondrial) catalyzes the oxidation of hydrogen sulfide, with the help of a quinone. Although the SQRDL precursor protein displays a molecular weight of 50 kDa, a prominent band at about 46 kDa is detected by western blot. This discrepancy reflects the cleavage of an N-terminal targeting sequence of 4 kDa during the import of the protein into the mitochondria. SQRDL is one of the enzymes involved in H2S signaling in a discrete population of neurons, oligodendrocytes, and endothelial cells.Specifications
SQRDL | |
Polyclonal | |
Unconjugated | |
SQOR | |
CGI 44, SQRDL | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
58472, 59010, 691966 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
0.14 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q9R112, Q9Y6N5 | |
Sqor | |
SQRDL Fusion Protein Ag10762 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |