Learn More
Uvex Sport NC 95179 Insole - Width 12
Comfortable climatic insole with antistatic properties for Uvex 1 Sport NC
19.85 EUR
Specifications
Color | Black |
---|---|
For Use With (Equipment) | Uvex 1 Sport NC |
Material | Microvelour |
Disposable | No |
Gender | Unisex |
Product Code | Brand | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Size | Price | Quantity & Availability | |||||
17267941
|
Uvex
9517935 |
35 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17277941
|
Uvex
9517936 |
36 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17287941
|
Uvex
9517937 |
37 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17297941
|
Uvex
9517938 |
38 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17207951
|
Uvex
9517939 |
39 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17217951
|
Uvex
9517940 |
40 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17227951
|
Uvex
9517941 |
41 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17237951
|
Uvex
9517942 |
42 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17247951
|
Uvex
9517943 |
43 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17257951
|
Uvex
9517944 |
44 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17267951
|
Uvex
9517945 |
45 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17277951
|
Uvex
9517946 |
46 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17287951
|
Uvex
9517947 |
47 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17297951
|
Uvex
9517948 |
48 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17207961
|
Uvex
9517949 |
49 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
17217961
|
Uvex
9517951 |
51 |
19.85 EUR
1 pair |
Estimated Shipment: 04-06-2024
Log in to see stock. |
|||||
Description
- Full-surface foot shock absorption
- Highly breathable and moisture-absorbent material
- Shock absorption zones support natural foot movement
- Skin compatibility and pleasant surface structure
- Supports foot arch
- Article No: 9517.9
- Sizes: 35 to 52
Specifications
Black | |
Microvelour | |
Unisex | |
Yes |
Uvex 1 Sport NC | |
No | |
Wide width | |
No |