Learn More
Manta Diagnostics™ Sponge Swab in NaCl
Includes sponge swab in NaCl 50 cm2, 60 sterile self seal sample bags and 60 pairs of sterile gloves
90.05 EUR - 92.60 EUR
Specifications
Applicator Material | Cellulose Sponge |
---|---|
Sterility | Sterile |
Color | Blue |
No. per Pack | 1 |
No. per Case | 60 |
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Includes | Price | Quantity & Availability | |||||
16604842
|
Manta Diagnostics™
T5 |
60 Swabs and 60 Grip Seal Bags, 60 pairs of Small gloves |
92.60 EUR
Pack of 60 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16694832
|
Manta Diagnostics™
T4 |
60 Swabs, 60 Grip Seal Bags, 60 pairs of Medium Gloves |
92.60 EUR
Pack of 60 |
Estimated Shipment: 13-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
16684832
|
Manta Diagnostics™
T3 |
60 Swabs, 60 Grip Seal Bags, 60 pairs of Large Gloves |
90.05 EUR
Pack of 60 |
Estimated Shipment: 05-08-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
Options of small, medium, or large glovesSpecifications
Cellulose Sponge | |
Blue | |
60 |
Sterile | |
1 |