Learn More
Spectrum™ Spectra/Gel™ Absorbent
Makes the concentration of protein solutions quick and efficient
Brand: Spectrum™ 292600
Additional Details : Weight : 2.72160kg
Description
- Material: Polyacrylate-polyalcohol
- Expands when it absorbs water
- Can be used with dialysis membranes with MWCOs from 0.1 to 300 kD
- Easy to remove
Specifications
Absorbent | |
Polyacrylate-Polyalcohol | |
Each |
0.1 to 300 kDa | |
White | |
For use with any MWCO dialysis membrane, high water absorbent capacity, convenient for sample concentration with Float-A-Lyzer™ G2 |
Length of time needed for concentration depends on the final volume, MWCO, surface-to-volume ratio of membrane tubing and amount of absorbent.