Learn More
Spectrum™ Bessman Tissue Pulverizers
Consists of a 2 component mortar with handles and a pestle, specifically designed for pulverizing 10 - 1000 mg of tissue
Brand:  Spectrum™ 189475
Additional Details : Weight : 1.00000kg
 View more versions of this product
Product Code. 15179843
  • 1125.00 EUR / Each
Estimated Shipment: 07-06-2024
Description
After chilling the mortar and pestle in liquid nitrogen, the included 28 cm lead hammer is used on the mortar to completely pulverize the inserted sample. The sample can then easily be transferred without touching or thawing by removing the mortar and inverting the pestle assembly over a test tube.
  • 3 sizes available: 10-50mg, 25-250mg and 100-1000mg of tissue
  • Small and Medium Pulverizers include 10 test tubes
  • Large Pulverizer includes one 90 mL culture jar
Specifications
Specifications
10 Test tubes, 11 in. Lead hammer
12.7mm
Designed for pulverizing 10 to 1000mg of tissue
0.5 in.
25 to 250mg
47.3mm
25 to 250mg
1.86 in.