Learn More
Labnet™ Spectrafuge™ 16M Brushless Laboratory Microcentrifuge
The Spectrafuge™ 16M Microcentrifuge is a high speed compact microcentrifuge for DNA applications.
Brand:  Labnet™ C0160-B-230V
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 11461997
  • 1290.00 EUR / Each
Estimated Shipment: 18-07-2024
to see stock.
Description
Description
  • The Spectrafuge™ 16M Microcentrifuge is the perfect high speed compact microcentrifuge. Producing a maximum of 16,000 x g, it is the ideal unit for DNA applications.
  • A brushless, maintenance free motor provides extremely fast acceleration and deceleration of the 18-place rotor and makes the Spectrafuge™ 16M one of the quietest microcentrifuges in its class.
  • Centrifuge Control Speed and time are set with the control knobs on the front panel of the unit. A 'Quick' button allows for momentary runs. A continuous flow of ambient air through the rotor chamber keeps the rotor and samples cool.
  • For applications requiring sub-ambient temperatures, the Spectrafuge™ 16M may be operated in a cold room.
  • The Spectrafuge™ 16M Microcentrifuge is supplied complete with an 18 place rotor for 1.5/2.0 mL tubes. Smaller tubes are accommodated by adapters (sold separately). The optional Strip-Spin™ rotor adapter holds two 8 x 0.2 mL strips, typically used for PCR.
    • Brushless, maintenance-free motor
    • Exceptionally quiet and cool operation
    • High maximum speed/g-force
    • Extremely small footprint
Specifications
Specifications
18 x 1.5/2.0 mL
1 to 30 min. or Continuous
1000 to 14,000 rpm
4.5 kg (10 lb.)
230 V
16,000 x g
4°C to 35°C
19.3 x 21 x 23 cm (7.6 x 8.25 x 8.9 in.)
50/60 Hz
Blue