Learn More
Spectrum Labs™ Spectra/Mesh™ Woven Nylon Filter Discs
Hydrophilic nylon filters with high strength for use in laboratory, industrial, or pharmaceutical applications.
Brand:  Spectrum Labs™ 145925
Additional Details : Weight : 0.50000kg
 View more versions of this product
Product Code. 15171996
  • 242.00 EUR / Pack of 10
Estimated Shipment: 12-06-2024
Description
  • Hydrophilic
  • Good resistance against corrosives
  • Resistant against pH 3 to 10
  • Sterilize via irradiation
Specifications
Specifications
Nylon
Laboratory, Industrial, Pharmaceutical
180°C
90 mm
5 μm
2%
100 μm