Learn More
Brannan™ Specific Gravity Hydrometer
Brand:  Brannan™ 10/924/0 + 49/040/0
Additional Details : Weight : 1.00000kg
Product Code. 15223997
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Specifications
1Each
0.001 S60/60°F
Steel Shot
Specific Gravity Hydrometer
270 mm
0.050 S60/60°F