Learn More
Bochem™ Spatula, Cement
Spatula for fillings, cements and pastes. Bochem™ Spatula, Cement has a sturdy, ridged handle.
Brand:  Bochem™ 3072
Additional Details : Weight : 0.10000kg
 View more versions of this product
Product Code. 10030751
  • 5.00 EUR / Each
Estimated Shipment: 31-05-2024
Description
  • Stainless steel construction.
  • Sturdy, ridged handle.
Specifications
Specifications
Round End, Pointed End
35 mm
Stainless Steel
Cements, fillings, pastes
6 mm
150 mm