Learn More
Labcold™ Sparkfree Freezers
For the safe storage of potentially flammable items. Labcold™ Sparkfree Freezers come in both chest and upright models.
Brand: Labcold™ RLCF0720
Additional Details : Weight : 1.00000kg
Description
- CFC and HCFC free
- Operator controlled defrost
- Alarm/temperature display can be fitted as an optional extra
- Chest freezers feature integral lid lock
- Locks available for all upright freezers as an additional extra
Specifications
215 L | |
1 Basket | |
RLCF0720 | |
860 mm |
-18°C to -25°C | |
Chest | |
730 mm | |
725 mm |