Learn More
Ebro Electronic™ SSX 210 Salt Meter Probe
Exchange probe for SSX 210, Probe SSX 210 with 150cm cable
Brand: Ebro Electronic™ 1341-5210
Additional Details : Weight : 0.80000kg
Description
Used to measure the salt content in fluid and semi-solid food products, such as meat, cold cuts, cheese, salads etc.Order Info
Type
Specifications
Probe | |
Not Calibrated | |
SPX 210 |
No Display | |
None |