missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SPARC Mouse anti-Human, Clone: 122511, R&D Systems™
Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB941-100
Additional Details : Weight : 0.00970kg
Description
SPARC Monoclonal specifically detects SPARC in Human samples. It is validated for ELISA, Immunohistochemistry, Intracellular Staining by Flow Cytometry, CyTOF-ready.Specifications
SPARC | |
Monoclonal | |
Unconjugated | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Basement-membrane protein 40, BM-40, ONcysteine-rich protein, osteonectin, Secreted protein acidic and rich in cysteine, secreted protein, acidic, cysteine-rich (osteonectin) | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6678 | |
Reconstitute at 0.5 mg/mL in sterile PBS. | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
IgG1 |
Immunohistochemistry, Flow Cytometry, CyTOF | |
122511 | |
Immunohistochemistry 8-25 ug/mL, Intracellular Staining by Flow Cytometry 0.25 ug/10^6 cells, CyTOF-ready | |
P09486 | |
SPARC | |
Mouse myeloma cell line NS0-derived recombinant human SPARC Ala18-Ile303 Accession # P09486 | |
100 μg | |
Primary | |
Detects human SPARC/Osteonectin in direct ELISAs. In direct ELISAs, no cross-reactivity with recombinant mouse SPARC/Osteonectin is observed. | |
Human | |
Purified |