Learn More
Fisherbrand™ Bonded 2mm thick Spacers
For use with notched and plain plates to ensure correct alignment and leak-free casting.
Brand:  Fisherbrand™ VS20S2
Product Code. 11864542
  • 18.50 EUR / Pack of 2
Estimated Shipment: 20-12-2024
Description
  • Accessory for use with Verti-GEL Maxi 2-Gel Vertical Gel Electrophoresis System
Specifications
Specifications
Verti-Gel Maxi 2-Gel Vertical Gel Electrophoresis System
Bonded 2 mm thick Spacers