missing translation for 'onlineSavingsMsg'
Learn More

Sodium Thiosulfate Pentahydrate, ≥99.5%, ReagentPlus™, Honeywell™

Catalog Number 15608120
Click to view available options
Quantity:
500 g
2.5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]
This item is not returnable. View return policy
10102-17-7
H10Na2O8S2
248.172
MFCD00149186
PODWXQQNRWNDGD-UHFFFAOYSA-L
sodium thiosulfate pentahydrate, ametox, sodium thiosulfate, pentahydrate, antichlor, tinver, disodium thiosulfate pentahydrate, unii-hx1032v43m, ccris 3952, thiosulfuric acid, disodium salt, pentahydrate, sodium hyposulfite pentahydrate
61475
CHEBI:32150
disodium;dioxido-oxo-sulfanylidene-$l^{6}-sulfane;pentahydrate
O.O.O.O.O.[O-]S(=O)(=S)[O-].[Na+].[Na+]

Reagent Grade,≥99.5%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point 48°C
Color Colorless
pH 6.0 to 7.5 (at 100.00g/L 25°C)
Quantity 500 g
Linear Formula Na2S2O3 · 5H2O
UN Number NONH for all modes of transport
Formula Weight 248.18g/mol
Percent Purity ≥99.5%
Odor Odorless
Packaging Poly bottle
Appearance Crystalline
Density 1.73g/cu.cm at 20°C
Chemical Name or Material Sodium thiosulfate pentahydrate
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'