missing translation for 'onlineSavingsMsg'
Learn More

Sodium Nitrate, Extra Pure, SLR, Fisher Chemical™

Catalog Number p-7073550
Click to view available options
Quantity:
500 g
1 kg
3 kg
This item is not returnable. View return policy
7631-99-4
NNaO3
84.99
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
24268
CHEBI:63005
[Na+].[O-][N+]([O-])=O
This item is not returnable. View return policy
7631-99-4
NNaO3
84.99
MFCD00011119
VWDWKYIASSYTQR-UHFFFAOYSA-N
sodium nitrate, chile saltpeter, nitrate of soda, cubic niter, etabisulfite, nitratine, soda niter, sodium saltpeter, nitric acid, sodium salt, sodium i nitrate 1:1
24268
CHEBI:63005
[Na+].[O-][N+]([O-])=O

Sodium Nitrate, Extra Pure, SLR

TRUSTED_SUSTAINABILITY
Boiling Point 380°C
Melting Point 306°C
Color White
pH 5.5 to 8
Physical Form Solid Crystalline
Quantity 500 g
IUPAC Name sodium;nitrate
Formula Weight 84.99g/mol
Packaging Plastic powder jar
compliance-icons compliance-icons
Product Identifier
  • Sodium nitrate
Signal Word
  • Warning
Hazard Category
  • Oxidising solid Category 3
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H272-May intensify fire; oxidiser.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P371+P380+P375-In case of major fire and large quantities: Evacuate area Fight fire remotely due to the risk of explosion.
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'