missing translation for 'onlineSavingsMsg'
Learn More

Sodium Metabisulfite, ≥99%, Reagent Grade, Honeywell™

Catalog Number 15658130
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Poly bottle
This item is not returnable. View return policy
7681-57-4
Na2O5S2
190.09
MFCD00167602
LDTLADDKFLAYJA-UHFFFAOYSA-L
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
656671
disodium (sulfinatooxy)sulfinate
[Na+].[Na+].[O-]S(=O)OS([O-])=O
This item is not returnable. View return policy
7681-57-4
Na2O5S2
190.09
MFCD00167602
LDTLADDKFLAYJA-UHFFFAOYSA-L
sodium metabisulfite, sodium pyrosulfite, sodium disulfite, disodium pyrosulfite, sodium metabisulphite, disodium disulfite, disodium disulphite, fertisilo, disodium metabisulfite, natrii disulfis
656671
disodium (sulfinatooxy)sulfinate
[Na+].[Na+].[O-]S(=O)OS([O-])=O

Reagent Grade,≥99%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY
Melting Point >150°C
Color Colorless
pH 3.5 to 5.0 (at 50.00g/L 20°C)
Physical Form Solid
Quantity 500 g
Linear Formula Na2S2O5
UN Number NONH for all modes of transport
Formula Weight 190.11g/mol
Percent Purity ≥99%
Odor Stinging
Packaging Poly bottle
Density 2.36g/cu.cm at 20°C
Chemical Name or Material Sodium metabisulfite
Show More Show Less
missing translation for 'provideContentCorrection'

missing translation for 'productTitle'